Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALB SIRNA Assays ›
Gene Symbol:
Alb
Gene Name:
albumin
Gene Aliases:
Alb1, Albza
Chromosome Location:
Chr.14: 19176275 - 19191793 on Build Rnor_6.0
UniGene ID:
Rn.202968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s127432
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Alb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_134326.2
NP_599153.2
9
1136
GenBank mRNA
BC085359.1
-
9
1136
7303856
FQ209382.1
-
5
584
FQ209565.1
-
2
169
FQ209580.1
-
2
121
FQ209635.1
-
3
346
FQ209651.1
-
3
376
FQ209693.1
-
2
171
FQ209695.1
-
5
570
FQ209713.1
-
1
46
FQ209714.1
-
2
239
FQ209721.1
-
3
357
FQ209825.1
-
2
162
FQ209907.1
-
5
605
FQ209957.1
-
4
517
FQ209968.1
-
2
164
FQ210022.1
-
3
359
FQ210057.1
-
5
535
FQ210064.1
-
1
28
FQ210111.1
-
3
388
FQ210171.1
-
3
357
FQ210205.1
-
1
49
FQ210297.1
-
2
204
FQ210310.1
-
7
808
FQ210316.1
-
3
357
FQ210347.1
-
2
138
FQ210361.1
-
2
277
FQ210399.1
-
2
102
FQ210406.1
-
3
336
FQ210411.1
-
2
268
FQ210445.1
-
1
42
FQ210540.1
-
2
86
FQ210546.1
-
3
384
FQ210564.1
-
2
121
FQ210640.1
-
3
341
FQ210724.1
-
6
660
FQ210749.1
-
1
60
FQ210803.1
-
5
606
FQ210850.1
-
4
452
FQ210851.1
-
4
483
FQ210858.1
-
6
690
FQ210877.1
-
5
551
FQ210946.1
-
1
45
FQ210995.1
-
3
346
FQ211001.1
-
2
176
FQ211010.1
-
4
494
FQ211036.1
-
3
389
FQ211096.1
-
3
357
FQ211119.1
-
1
46
FQ211129.1
-
3
330
FQ211183.1
-
2
108
FQ211196.1
-
3
317
FQ211218.1
-
6
764
FQ211226.1
-
1
39
FQ211255.1
-
5
584
FQ211259.1
-
5
593
FQ211355.1
-
2
181
FQ211376.1
-
1
40
FQ211378.1
-
3
357
FQ211445.1
-
2
261
FQ218081.1
-
2
165
FQ218137.1
-
3
361
FQ218216.1
-
4
460
FQ218268.1
-
1
57
FQ218283.1
-
2
254
FQ218310.1
-
1
35
FQ218331.1
-
1
35
FQ218371.1
-
1
67
FQ218379.1
-
1
43
FQ218412.1
-
2
168
FQ218458.1
-
5
600
FQ218497.1
-
1
28
FQ218566.1
-
2
267
FQ218682.1
-
3
325
FQ218739.1
-
4
505
FQ218757.1
-
1
46
FQ218799.1
-
2
90
FQ218806.1
-
1
42
FQ218815.1
-
4
470
FQ218867.1
-
2
286
FQ218989.1
-
2
190
FQ219017.1
-
2
141
FQ219210.1
-
5
555
FQ219231.1
-
2
182
FQ219260.1
-
3
326
FQ219293.1
-
5
584
FQ219382.1
-
2
271
FQ219447.1
-
5
577
FQ219470.1
-
4
455
FQ219477.1
-
2
197
FQ219514.1
-
3
383
FQ219650.1
-
3
353
FQ219670.1
-
5
634
FQ219679.1
-
3
346
FQ219682.1
-
3
328
FQ219706.1
-
1
66
FQ219775.1
-
2
240
V01222.1
-
9
1121
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009654, AK131891, AJ011413, BC024643, AJ457860, and others.
Gene Ontology Categories:
Function(s)
Process(es)
retina homeostasis
transport
response to nutrient
cellular response to starvation
response to organic substance
hemolysis by symbiont of host erythrocytes
vasodilation
negative regulation of apoptotic process
positive regulation of circadian sleep/wake cycle, non-REM sleep
response to mercury ion
maintenance of mitochondrion location
response to platinum ion
DNA binding
fatty acid binding
drug binding
zinc ion binding
toxic substance binding
oxygen binding
enzyme binding
pyridoxal phosphate binding
chaperone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix