Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALB SIRNA Assays ›
          Gene Symbol:
          Alb
          Gene Name:
          albumin
          Gene Aliases:
          Alb1, Albza
          Chromosome Location:
          Chr.14: 19176275 - 19191793 on Build Rnor_6.0
          UniGene ID:
          Rn.202968
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s127434

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Alb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_134326.2 NP_599153.2 10 1245
          GenBank mRNA BC085359.1 - 10 1245 7303856
          FQ209382.1 - 6 693
          FQ209565.1 - 3 278
          FQ209580.1 - 3 230
          FQ209595.1 - 2 173
          FQ209635.1 - 4 456
          FQ209651.1 - 4 485
          FQ209693.1 - 3 280
          FQ209695.1 - 6 679
          FQ209713.1 - 2 155
          FQ209714.1 - 3 348
          FQ209721.1 - 4 466
          FQ209750.1 - 1 24
          FQ209825.1 - 3 271
          FQ209907.1 - 6 714
          FQ209910.1 - 1 18
          FQ209930.1 - 1 17
          FQ209957.1 - 5 626
          FQ209968.1 - 3 273
          FQ210022.1 - 4 468
          FQ210057.1 - 6 644
          FQ210064.1 - 2 137
          FQ210111.1 - 4 497
          FQ210134.1 - 2 60
          FQ210171.1 - 4 466
          FQ210205.1 - 2 158
          FQ210297.1 - 3 313
          FQ210310.1 - 8 917
          FQ210316.1 - 4 466
          FQ210361.1 - 3 386
          FQ210399.1 - 3 211
          FQ210406.1 - 4 445
          FQ210411.1 - 3 377
          FQ210445.1 - 2 151
          FQ210540.1 - 3 195
          FQ210546.1 - 4 493
          FQ210564.1 - 3 230
          FQ210585.1 - 2 101
          FQ210640.1 - 4 450
          FQ210749.1 - 2 169
          FQ210803.1 - 6 717
          FQ210850.1 - 5 561
          FQ210851.1 - 5 592
          FQ210858.1 - 7 800
          FQ210872.1 - 1 50
          FQ210877.1 - 6 660
          FQ210926.1 - 2 63
          FQ210946.1 - 2 154
          FQ210954.1 - 2 233
          FQ210973.1 - 1 24
          FQ210995.1 - 4 455
          FQ211001.1 - 3 285
          FQ211010.1 - 5 603
          FQ211021.1 - 1 31
          FQ211036.1 - 4 498
          FQ211096.1 - 4 466
          FQ211119.1 - 2 155
          FQ211129.1 - 4 439
          FQ211131.1 - 1 30
          FQ211157.1 - 2 109
          FQ211196.1 - 4 426
          FQ211226.1 - 2 148
          FQ211255.1 - 6 693
          FQ211259.1 - 6 702
          FQ211320.1 - 4 497
          FQ211355.1 - 3 290
          FQ211376.1 - 2 149
          FQ211378.1 - 4 466
          FQ211445.1 - 3 370
          FQ211464.1 - 2 70
          FQ218081.1 - 3 274
          FQ218137.1 - 4 470
          FQ218216.1 - 5 569
          FQ218268.1 - 2 166
          FQ218283.1 - 3 363
          FQ218304.1 - 2 110
          FQ218310.1 - 2 144
          FQ218331.1 - 2 144
          FQ218336.1 - 2 74
          FQ218371.1 - 2 176
          FQ218379.1 - 2 152
          FQ218412.1 - 3 277
          FQ218458.1 - 6 709
          FQ218497.1 - 2 137
          FQ218541.1 - 2 107
          FQ218566.1 - 3 376
          FQ218682.1 - 4 434
          FQ218739.1 - 5 614
          FQ218757.1 - 2 155
          FQ218799.1 - 3 199
          FQ218806.1 - 2 151
          FQ218815.1 - 5 579
          FQ218867.1 - 3 395
          FQ218944.1 - 3 308
          FQ218989.1 - 3 299
          FQ219017.1 - 3 250
          FQ219210.1 - 6 664
          FQ219231.1 - 3 291
          FQ219260.1 - 4 435
          FQ219293.1 - 6 693
          FQ219382.1 - 3 380
          FQ219442.1 - 4 463
          FQ219470.1 - 5 564
          FQ219477.1 - 3 306
          FQ219514.1 - 4 492
          FQ219650.1 - 4 462
          FQ219670.1 - 6 743
          FQ219679.1 - 4 455
          FQ219682.1 - 4 437
          FQ219706.1 - 2 175
          FQ219734.1 - 8 896
          FQ219775.1 - 3 349
          FQ224096.1 - 3 349
          V01222.1 - 10 1230

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          transport
          response to nutrient
          cellular response to starvation
          response to organic substance
          hemolysis by symbiont of host erythrocytes
          vasodilation
          negative regulation of apoptotic process
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          response to mercury ion
          maintenance of mitochondrion location
          response to platinum ion
          DNA binding
          fatty acid binding
          drug binding
          zinc ion binding
          toxic substance binding
          oxygen binding
          enzyme binding
          pyridoxal phosphate binding
          chaperone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix