Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B2M SIRNA Assays ›
          Gene Symbol:
          B2m
          Gene Name:
          beta-2 microglobulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 114087287 - 114093311 on Build Rnor_6.0
          UniGene ID:
          Rn.1868
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s127498

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B2m
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012512.2 NP_036644.1 2 313
          GenBank mRNA FM032790.1 - 2 313
          FQ210051.1 - 2 326
          FQ210252.1 - 2 327
          FQ210804.1 - 2 325
          FQ210906.1 - 2 326
          FQ211296.1 - 2 326
          FQ211419.1 - 2 325
          FQ212410.1 - 2 327
          FQ217926.1 - 2 324
          FQ218141.1 - 2 326
          FQ218251.1 - 2 326
          FQ218298.1 - 2 325
          FQ219157.1 - 2 328
          FQ219455.1 - 875
          FQ220256.1 - 2 322
          FQ221268.1 - 2 326
          FQ221287.1 - 2 322
          FQ221985.1 - 2 325
          FQ222352.1 - 2 323
          FQ225010.1 - 2 326
          FQ225051.1 - 2 330
          FQ225087.1 - 2 325
          FQ225088.1 - 2 327
          FQ225121.1 - 2 325
          FQ225141.1 - 2 325
          FQ225219.1 - 2 324
          FQ225292.1 - 2 326
          FQ225550.1 - 2 325
          FQ225647.1 - 2 325
          FQ225686.1 - 2 328
          FQ225690.1 - 2 467
          FQ225847.1 - 2 325
          FQ225912.1 - 2 325
          FQ226004.1 - 2 326
          FQ226258.1 - 2 328
          FQ226642.1 - 2 327
          FQ226723.1 - 2 325
          FQ227149.1 - 2 325
          FQ227291.1 - 2 331
          FQ227327.1 - 2 326
          FQ227454.1 - 2 326
          FQ227468.1 - 2 324
          FQ227656.1 - 2 326
          FQ227950.1 - 2 324
          FQ227964.1 - 2 325
          FQ227995.1 - 2 321
          FQ228065.1 - 2 327
          FQ228188.1 - 2 326
          FQ228427.1 - 2 327
          FQ229482.1 - 2 318
          FQ229718.1 - 2 325
          FQ230202.1 - 2 326
          FQ230402.1 - 2 326
          FQ230977.1 - 2 325
          FQ231135.1 - 2 326
          FQ231182.1 - 2 326
          FQ231367.1 - 2 325
          FQ231378.1 - 2 325
          FQ231524.1 - 2 325
          FQ231602.1 - 2 326
          FQ231718.1 - 2 325
          FQ231856.1 - 2 325
          FQ231894.1 - 2 325
          FQ232233.1 - 2 326
          FQ232294.1 - 2 326
          FQ232460.1 - 2 325
          FQ232758.1 - 2 325
          FQ232818.1 - 2 325
          FQ232819.1 - 2 325
          FQ232937.1 - 2 325
          FQ232943.1 - 2 327
          FQ232957.1 - 2 323
          FQ233220.1 - 2 326
          FQ233240.1 - 2 326
          FQ233290.1 - 2 325
          FQ233447.1 - 2 326
          FQ233505.1 - 2 325
          FQ233544.1 - 2 325
          FQ233567.1 - 2 330
          FQ233757.1 - 2 325
          FQ233797.1 - 2 324
          FQ233849.1 - 2 325
          FQ234348.1 - 2 326
          FQ234349.1 - 2 325
          FQ234524.1 - 2 323
          FQ234581.1 - 2 323
          FQ234840.1 - 2 325
          FQ234885.1 - 2 325
          FQ235105.1 - 2 325
          FQ235149.1 - 2 323
          FQ235229.1 - 2 325
          X16956.1 - 1 102
          Y00441.1 - 2 300
          Y08531.1 - 2 305

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          T cell differentiation in thymus
          protein refolding
          response to drug
          response to cadmium ion
          iron ion homeostasis
          molecular_function
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-bff4f7d6d-tmrcx:80/100.66.135.11:80.
          git-commit: 683fd26f943b2dde6a08880618459b58d20c3896
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline