Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B2M SIRNA Assays ›
          Gene Symbol:
          B2m
          Gene Name:
          beta-2 microglobulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 114087287 - 114093311 on Build Rnor_6.0
          UniGene ID:
          Rn.1868
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s127500

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B2m
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012512.2 NP_036644.1 2 219
          GenBank mRNA FM032790.1 - 2 219
          FQ210051.1 - 2 232
          FQ210252.1 - 2 233
          FQ210804.1 - 2 231
          FQ210906.1 - 2 232
          FQ211296.1 - 2 232
          FQ211419.1 - 2 231
          FQ212410.1 - 2 233
          FQ217926.1 - 2 230
          FQ218141.1 - 2 232
          FQ218251.1 - 2 232
          FQ218298.1 - 2 231
          FQ219157.1 - 2 234
          FQ219455.1 - 781
          FQ221268.1 - 2 232
          FQ221287.1 - 2 228
          FQ221985.1 - 2 231
          FQ222352.1 - 2 229
          FQ225010.1 - 2 232
          FQ225051.1 - 2 236
          FQ225087.1 - 2 231
          FQ225088.1 - 2 233
          FQ225121.1 - 2 231
          FQ225141.1 - 2 231
          FQ225219.1 - 2 230
          FQ225292.1 - 2 232
          FQ225550.1 - 2 231
          FQ225647.1 - 2 231
          FQ225686.1 - 2 234
          FQ225690.1 - 2 373
          FQ225847.1 - 2 231
          FQ225912.1 - 2 231
          FQ226004.1 - 2 232
          FQ226258.1 - 2 234
          FQ226642.1 - 2 233
          FQ226723.1 - 2 231
          FQ227149.1 - 2 231
          FQ227291.1 - 2 237
          FQ227454.1 - 2 232
          FQ227468.1 - 2 230
          FQ227656.1 - 2 232
          FQ227700.1 - 2 232
          FQ227950.1 - 2 230
          FQ227964.1 - 2 231
          FQ227995.1 - 2 227
          FQ228065.1 - 2 233
          FQ228188.1 - 2 232
          FQ229482.1 - 2 224
          FQ229718.1 - 2 231
          FQ230202.1 - 2 232
          FQ230402.1 - 2 232
          FQ230977.1 - 2 231
          FQ231135.1 - 2 232
          FQ231182.1 - 2 232
          FQ231367.1 - 2 231
          FQ231378.1 - 2 231
          FQ231524.1 - 2 231
          FQ231602.1 - 2 232
          FQ231718.1 - 2 231
          FQ231856.1 - 2 231
          FQ231894.1 - 2 231
          FQ232233.1 - 2 232
          FQ232294.1 - 2 232
          FQ232460.1 - 2 231
          FQ232758.1 - 2 231
          FQ232818.1 - 2 231
          FQ232819.1 - 2 231
          FQ232937.1 - 2 231
          FQ232943.1 - 2 233
          FQ232957.1 - 2 229
          FQ233220.1 - 2 232
          FQ233240.1 - 2 232
          FQ233290.1 - 2 231
          FQ233447.1 - 2 232
          FQ233505.1 - 2 231
          FQ233544.1 - 2 231
          FQ233567.1 - 2 236
          FQ233757.1 - 2 231
          FQ233797.1 - 2 230
          FQ233849.1 - 2 231
          FQ234348.1 - 2 232
          FQ234349.1 - 2 231
          FQ234524.1 - 2 229
          FQ234581.1 - 2 229
          FQ234840.1 - 2 231
          FQ234885.1 - 2 231
          FQ235149.1 - 2 229
          FQ235229.1 - 2 231
          Y00441.1 - 2 206
          Y08531.1 - 2 211

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          T cell differentiation in thymus
          protein refolding
          response to drug
          response to cadmium ion
          iron ion homeostasis
          molecular_function
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-bff4f7d6d-tmrcx:80/100.66.135.11:80.
          git-commit: 683fd26f943b2dde6a08880618459b58d20c3896
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline