Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERBB2 SIRNA Assays ›
          Gene Symbol:
          Erbb2 LOC102552659
          Gene Name:
          v-erb-b2 avian erythroblastic leukemia viral oncogene 2
          receptor tyrosine-protein kinase erbB-2-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.10: 86367596 - 86391728 on Build Rnor_6.0
          Chromosome Location:
          Chr.10: 84838237 - 84845803 on Build Rnor_6.0
          UniGene ID:
          Rn.93966
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s127711

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Erbb2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017003.2 NP_058699.2 26, 27 3533
          GenBank mRNA AY116182.1 - 26, 27 3533
          BC061863.1 - 26, 27 3605 5597835
          X03362.1 - 26, 27 3447
          Gene Symbol: LOC102552659
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006247245.2 XP_006247307.2 13, 14 1796

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          signal transduction
          cell surface receptor signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          nervous system development
          central nervous system development
          peripheral nervous system development
          heart development
          skeletal muscle tissue development
          neuromuscular junction development
          motor neuron axon guidance
          positive regulation of cell proliferation
          glial cell differentiation
          phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          signal transduction by protein phosphorylation
          positive regulation of cell growth
          response to progesterone
          regulation of microtubule-based process
          negative regulation of immature T cell proliferation in thymus
          wound healing
          regulation of cell proliferation
          response to drug
          myelination
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          tongue development
          estrous cycle
          regulation of cell differentiation
          positive regulation of translation
          positive regulation of cell adhesion
          positive regulation of transcription from RNA polymerase I promoter
          positive regulation of transcription from RNA polymerase III promoter
          positive regulation of Ras protein signal transduction
          protein autophosphorylation
          sympathetic nervous system development
          response to axon injury
          oligodendrocyte differentiation
          positive regulation of epithelial cell proliferation
          mammary gland involution
          regulation of ERK1 and ERK2 cascade
          RNA polymerase I core binding
          glycoprotein binding
          DNA binding
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          transmembrane signaling receptor activity
          protein binding
          ATP binding
          protein C-terminus binding
          growth factor binding
          protein phosphatase binding
          ubiquitin protein ligase binding
          identical protein binding
          protein heterodimerization activity
          Hsp90 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0