Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GRIN2A SIRNA Assays ›
          Gene Symbol:
          Grin2a
          Gene Name:
          glutamate receptor, ionotropic, N-methyl D-aspartate 2A
          Gene Aliases:
          GluN2A, NMDAR2A, NR2A
          Chromosome Location:
          Chr.10: 5707806 - 6119897 on Build Rnor_6.0
          UniGene ID:
          Rn.9710
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s127809

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Grin2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012573.3 NP_036705.3 13 4412
          GenBank mRNA D13211.1 - 12 4205
          M91561.1 - 13 4412

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          action potential
          startle response
          response to amphetamine
          cation transport
          calcium ion transport
          chemical synaptic transmission
          learning or memory
          learning
          memory
          protein localization
          visual learning
          response to light stimulus
          response to wounding
          response to carbohydrate
          response to manganese ion
          positive regulation of cell death
          response to organic cyclic compound
          response to amine
          sensory perception of pain
          spinal cord development
          hippocampus development
          cerebral cortex development
          neurogenesis
          sleep
          directional locomotion
          ion transmembrane transport
          regulation of ion transmembrane transport
          ionotropic glutamate receptor signaling pathway
          synaptic transmission, glutamatergic
          locomotion
          negative regulation of protein catabolic process
          response to cocaine
          regulation of membrane potential
          dopamine metabolic process
          serotonin metabolic process
          response to drug
          positive regulation of apoptotic process
          response to ethanol
          regulation of synaptic plasticity
          regulation of long-term neuronal synaptic plasticity
          rhythmic process
          modulation of synaptic transmission
          detection of mechanical stimulus involved in sensory perception of pain
          protein tetramerization
          response to calcium ion
          response to methylmercury
          response to other organism
          regulation of sensory perception of pain
          regulation of postsynaptic membrane potential
          excitatory postsynaptic potential
          long-term synaptic potentiation
          response to ammonium ion
          response to fungicide
          calcium ion transmembrane transport
          cellular response to amino acid stimulus
          cellular response to magnesium ion
          cellular response to manganese ion
          cellular response to zinc ion
          cellular response to dsRNA
          cellular response to growth factor stimulus
          cellular response to lipid
          cation transmembrane transport
          positive regulation of excitatory postsynaptic potential
          ionotropic glutamate receptor activity
          NMDA glutamate receptor activity
          receptor binding
          extracellular-glutamate-gated ion channel activity
          cation channel activity
          calcium channel activity
          protein binding
          zinc ion binding
          glutamate binding
          protein kinase binding
          voltage-gated cation channel activity
          protein complex binding
          glutamate receptor binding
          neurotransmitter binding
          protein heterodimerization activity
          protein dimerization activity
          cell adhesion molecule binding
          ATPase binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0