Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022797.1 | NP_073634.1 | 10 | 2927 | ||
XM_008759316.1 | XP_008757538.1 | 9 | 2632 | |||
XM_008759317.1 | XP_008757539.1 | 9 | 2632 | |||
GenBank mRNA | D13213.1 | - | 9 | 2221 | ||
L31611.1 | - | 9 | 2202 | |||
L31612.1 | - | 10 | 2927 | |||
U08260.1 | - | 10 | 2273 |