Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL1A SIRNA Assays ›
          Gene Symbol:
          Il1a
          Gene Name:
          interleukin 1 alpha
          Gene Aliases:
          IL-1 alpha
          Chromosome Location:
          Chr.3: 121824712 - 121836122 on Build Rnor_6.0
          UniGene ID:
          Rn.12300
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s127940

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Il1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017019.1 NP_058715.1 2, 3 105
          GenBank mRNA AJ245642.1 - 2, 3 69
          AJ245643.1 - 1, 2 44
          D00403.1 - 2, 3 105

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          fever generation
          response to hypoxia
          connective tissue replacement involved in inflammatory response wound healing
          acute inflammatory response
          inflammatory response
          immune response
          spermatogenesis
          heart development
          negative regulation of cell proliferation
          response to heat
          response to organic substance
          response to ozone
          response to organonitrogen compound
          response to gamma radiation
          positive regulation of vascular endothelial growth factor production
          positive regulation of steroid biosynthetic process
          response to organic cyclic compound
          cytokine-mediated signaling pathway
          keratinization
          positive regulation of prostaglandin secretion
          response to lipopolysaccharide
          positive regulation of interleukin-6 production
          positive regulation of stress-activated MAPK cascade
          response to L-ascorbic acid
          cellular response to heat
          ectopic germ cell programmed cell death
          wound healing
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of angiogenesis
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          response to copper ion
          positive regulation of cytokine secretion
          positive regulation of cell division
          regulation of sensory perception of pain
          positive regulation of ERK1 and ERK2 cascade
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of neutrophil migration
          cytokine activity
          interleukin-1 receptor binding
          copper ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix