Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017024.2 | NP_058720.2 | 1 | 133 | ||
GenBank mRNA | BC091155.1 | - | 1 | 133 | 7370790 | |
FQ209624.1 | - | 1 | 148 | |||
FQ218245.1 | - | 1 | 148 | |||
FQ218883.1 | - | 1 | 151 | |||
U62803.1 | - | 1 | 136 | |||
X54096.1 | - | 1 | 147 |