Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pthlh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012636.1 | NP_036768.1 | 411 | |||
GenBank mRNA | M21967.1 | - | 337 | |||
M31603.1 | - | 411 |