Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012638.1 | NP_036770.1 | 20 | 2451 | ||
XM_006230678.2 | XP_006230740.1 | 20 | 2491 | |||
GenBank mRNA | BC161897.1 | - | 20 | 2446 | 8366912 | |
FQ217890.1 | - | 7 | 731 | |||
L10669.1 | - | 20 | 2402 | |||
X03032.1 | - | 3 | 120 |