Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPARC SIRNA Assays ›
Gene Symbol:
Sparc
Gene Name:
secreted protein, acidic, cysteine-rich (osteonectin)
Gene Aliases:
-
Chromosome Location:
Chr.10: 40742390 - 40764222 on Build Rnor_6.0
UniGene ID:
Rn.234316
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s128433
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sparc
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012656.1
NP_036788.1
10
836
XM_008767663.1
XP_008765885.1
9
914
GenBank mRNA
BC061777.1
-
9
851
5621666
D28875.1
-
9
853
FQ212364.1
-
9
905
FQ212499.1
-
9
871
FQ213815.1
-
9
870
FQ213945.1
-
9
870
FQ214222.1
-
9
868
FQ214492.1
-
9
870
FQ214649.1
-
9
871
FQ214673.1
-
9
870
FQ215195.1
-
9
869
FQ215581.1
-
9
871
FQ215622.1
-
9
872
FQ215818.1
-
9
881
FQ215935.1
-
9
870
FQ216823.1
-
9
872
FQ217546.1
-
9
871
FQ219866.1
-
9
852
FQ219932.1
-
10
852
FQ219983.1
-
9
870
FQ220003.1
-
9
870
FQ220043.1
-
9
868
FQ220046.1
-
9
870
FQ220186.1
-
9
868
FQ220279.1
-
7
694
FQ220287.1
-
9
872
FQ220443.1
-
9
852
FQ220507.1
-
9
878
FQ220547.1
-
9
870
FQ220644.1
-
9
870
FQ220827.1
-
9
870
FQ220880.1
-
9
870
FQ220901.1
-
9
870
FQ222224.1
-
9
870
FQ223025.1
-
9
870
FQ224996.1
-
8
741
FQ225033.1
-
9
872
FQ225092.1
-
9
866
FQ225200.1
-
9
870
FQ225220.1
-
9
849
FQ225486.1
-
9
852
FQ225609.1
-
9
870
FQ225618.1
-
9
868
FQ225644.1
-
5
537
FQ225678.1
-
9
870
FQ225688.1
-
9
870
FQ225694.1
-
9
872
FQ225722.1
-
9
869
FQ225884.1
-
9
868
FQ225913.1
-
9
870
FQ225914.1
-
9
872
FQ225965.1
-
9
870
FQ226299.1
-
9
852
FQ226354.1
-
9
870
FQ226360.1
-
9
870
FQ226584.1
-
9
870
FQ226599.1
-
9
871
FQ226651.1
-
9
893
FQ226673.1
-
9
852
FQ226731.1
-
9
870
FQ226746.1
-
9
868
FQ227279.1
-
9
871
FQ227628.1
-
9
879
FQ227644.1
-
9
870
FQ227708.1
-
9
871
FQ227725.1
-
9
871
FQ227727.1
-
9
850
FQ227749.1
-
9
870
FQ228032.1
-
9
850
FQ228597.1
-
9
852
FQ229172.1
-
9
870
FQ229199.1
-
9
870
FQ229230.1
-
9
870
FQ229283.1
-
9
852
FQ229299.1
-
9
853
FQ229315.1
-
9
868
FQ229316.1
-
9
870
FQ230029.1
-
9
870
FQ230086.1
-
9
868
FQ230210.1
-
9
871
FQ230381.1
-
9
870
FQ230595.1
-
9
852
FQ230619.1
-
9
871
FQ230667.1
-
9
871
FQ230705.1
-
9
868
FQ230778.1
-
9
870
FQ230864.1
-
9
870
FQ230993.1
-
9
870
FQ231067.1
-
9
870
FQ231166.1
-
9
869
FQ231444.1
-
9
869
FQ231523.1
-
9
870
FQ231543.1
-
9
870
FQ231632.1
-
9
870
FQ231753.1
-
9
867
FQ231815.1
-
9
852
FQ231918.1
-
9
852
FQ231975.1
-
9
852
FQ232042.1
-
9
869
FQ232191.1
-
9
870
FQ232201.1
-
9
870
FQ232223.1
-
9
870
FQ232271.1
-
9
867
FQ232364.1
-
9
872
FQ232445.1
-
9
868
FQ232464.1
-
9
870
FQ232710.1
-
9
868
FQ232838.1
-
9
870
FQ232847.1
-
9
870
FQ232862.1
-
9
868
FQ232869.1
-
9
870
FQ232965.1
-
9
870
FQ232977.1
-
9
870
FQ232986.1
-
9
870
FQ233178.1
-
9
870
FQ233283.1
-
9
849
FQ233669.1
-
9
870
FQ233698.1
-
9
852
FQ233777.1
-
9
868
FQ233810.1
-
9
872
FQ233845.1
-
9
870
FQ233850.1
-
9
871
FQ233926.1
-
9
870
FQ234037.1
-
9
870
FQ234069.1
-
9
870
FQ234108.1
-
9
870
FQ234199.1
-
9
870
FQ234410.1
-
9
852
FQ234418.1
-
9
870
FQ234523.1
-
9
857
FQ234589.1
-
9
868
FQ234657.1
-
9
852
FQ234685.1
-
9
870
FQ234766.1
-
9
868
FQ235008.1
-
9
872
FQ235038.1
-
9
871
FQ235049.1
-
9
852
Y13714.1
-
10
836
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
ossification
negative regulation of endothelial cell proliferation
signal transduction
heart development
response to gravity
response to lead ion
positive regulation of endothelial cell migration
negative regulation of angiogenesis
regulation of cell morphogenesis
lung development
response to lipopolysaccharide
response to L-ascorbic acid
response to cytokine
wound healing
regulation of cell proliferation
response to peptide hormone
response to ethanol
response to cadmium ion
inner ear development
response to glucocorticoid
response to cAMP
response to calcium ion
bone development
cellular response to growth factor stimulus
calcium ion binding
collagen binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0