Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173290.1 | NP_775412.1 | 4 | 1009 | ||
GenBank mRNA | AB080317.1 | - | 4 | 1009 |