Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP7A SIRNA Assays ›
          Gene Symbol:
          Atp7a
          Gene Name:
          ATPase, Cu++ transporting, alpha polypeptide
          Gene Aliases:
          Mnk
          Chromosome Location:
          Chr.X: 77076087 - 77193644 on Build Rnor_6.0
          UniGene ID:
          Rn.10554
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s128719

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Atp7a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_052803.2 NP_434690.1 7 1922
          XM_006256995.2 XP_006257057.1 8 2089
          XM_006256997.2 XP_006257059.1 8 2086
          XM_008773334.1 XP_008771556.1 8 2030
          GenBank mRNA U59245.1 - 7 1927

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_009726, NM_001109757, U03434, AB007134, BC141395, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          in utero embryonic development
          release of cytochrome c from mitochondria
          blood vessel remodeling
          regulation of oxidative phosphorylation
          tryptophan metabolic process
          tyrosine metabolic process
          catecholamine metabolic process
          copper ion transport
          cellular copper ion homeostasis
          mitochondrion organization
          lactation
          locomotory behavior
          response to iron(III) ion
          response to zinc ion
          detoxification of copper ion
          regulation of gene expression
          copper ion import
          plasma membrane copper ion transport
          peptidyl-lysine modification
          removal of superoxide radicals
          cerebellar Purkinje cell differentiation
          pyramidal neuron development
          central nervous system neuron development
          metal ion transport
          extracellular matrix organization
          collagen fibril organization
          hair follicle morphogenesis
          hindlimb morphogenesis
          T-helper cell differentiation
          epinephrine metabolic process
          norepinephrine metabolic process
          dopamine metabolic process
          norepinephrine biosynthetic process
          serotonin metabolic process
          positive regulation of catalytic activity
          negative regulation of catalytic activity
          pigmentation
          negative regulation of neuron apoptotic process
          skin development
          ATP metabolic process
          response to copper ion
          elastic fiber assembly
          lung alveolus development
          neuron projection morphogenesis
          dendrite morphogenesis
          cartilage development
          positive regulation of oxidoreductase activity
          elastin biosynthetic process
          copper ion export
          molecular_function
          copper-exporting ATPase activity
          copper ion transmembrane transporter activity
          copper ion binding
          ATP binding
          superoxide dismutase copper chaperone activity
          copper-dependent protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
          git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix