Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRD5A1 SIRNA Assays ›
          Gene Symbol:
          Srd5a1
          Gene Name:
          steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1)
          Gene Aliases:
          S5AR 1
          Chromosome Location:
          Chr.1: 36320504 - 36354694 on Build Rnor_6.0
          UniGene ID:
          Rn.4620
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s128742

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Srd5a1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017070.3 NP_058766.2 717
          GenBank mRNA BC127454.1 - 718 7313862
          EF091857.1 - 203
          J05035.1 - 570
          S81448.1 - 590

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          urogenital system development
          liver development
          steroid biosynthetic process
          androgen biosynthetic process
          androgen metabolic process
          male gonad development
          cellular response to starvation
          response to organic cyclic compound
          response to muscle activity
          diterpenoid metabolic process
          spinal cord development
          hippocampus development
          thalamus development
          hypothalamus development
          pituitary gland development
          cerebral cortex development
          cell differentiation
          male genitalia development
          female genitalia development
          response to follicle-stimulating hormone
          response to estradiol
          cellular response to insulin stimulus
          response to testosterone
          serotonin metabolic process
          progesterone metabolic process
          response to drug
          circadian sleep/wake cycle, REM sleep
          response to estrogen
          male sex differentiation
          oxidation-reduction process
          bone development
          response to growth hormone
          response to fungicide
          cellular response to cAMP
          cellular response to growth factor stimulus
          cellular response to estradiol stimulus
          cellular response to testosterone stimulus
          cellular response to organic cyclic compound
          cellular response to dexamethasone stimulus
          cellular response to epinephrine stimulus
          3-oxo-5-alpha-steroid 4-dehydrogenase activity
          amide binding
          cholestenone 5-alpha-reductase activity
          NADPH binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0