Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270575.1 | NP_001257504.1 | 6 | 634 | ||
NM_001270576.1 | NP_001257505.1 | 6 | 546 | |||
NM_030991.3 | NP_112253.1 | 6 | 634 | |||
GenBank mRNA | AB003991.1 | - | 425 | |||
AB003992.1 | - | 5 | 425 | |||
AF245227.1 | - | 6 | 633 | |||
BC087699.1 | - | 6 | 589 | 7314318 | ||
DY315301.1 | - | 6 | 566 | |||
FM033263.1 | - | 6 | 546 | |||
FQ212040.1 | - | 6 | 605 | |||
FQ213923.1 | - | 6 | 605 | |||
FQ214162.1 | - | 6 | 634 | |||
U56261.1 | - | 398 | ||||
U56262.1 | - | 5 | 398 |