Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022178.1 | NP_071514.1 | 186 | |||
GenBank mRNA | AB035736.1 | - | 186 |