Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STAT3 SIRNA Assays ›
          Gene Symbol:
          Stat3
          Gene Name:
          signal transducer and activator of transcription 3 (acute-phase response factor)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 88790401 - 88842257 on Build Rnor_6.0
          UniGene ID:
          Rn.10247
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s129047

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Stat3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012747.2 NP_036879.1 13 1430
          XM_006247257.2 XP_006247319.1 13 1454
          XM_006247258.2 XP_006247320.1 13 1454
          XM_006247259.2 XP_006247321.1 13 1454
          GenBank mRNA BC087025.1 - 13 1430 7110655
          X91810.1 - 12 1224

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_213659, NM_213660, NM_011486, BC019168, AK153005, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          eye photoreceptor cell differentiation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          protein import into nucleus
          acute-phase response
          JAK-STAT cascade
          aging
          cell proliferation
          negative regulation of cell proliferation
          response to organic substance
          negative regulation of hydrogen peroxide biosynthetic process
          response to organic cyclic compound
          phosphorylation
          stem cell population maintenance
          sexual reproduction
          intracellular receptor signaling pathway
          response to estradiol
          cellular response to hormone stimulus
          response to cytokine
          regulation of multicellular organism growth
          response to drug
          glucose homeostasis
          eating behavior
          response to peptide hormone
          response to ethanol
          positive regulation of Notch signaling pathway
          negative regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of mitochondrial membrane permeability
          astrocyte differentiation
          radial glial cell differentiation
          growth hormone receptor signaling pathway
          JAK-STAT cascade involved in growth hormone signaling pathway
          negative regulation of cell death
          interleukin-6-mediated signaling pathway
          negative regulation of neuron death
          positive regulation of growth factor dependent skeletal muscle satellite cell proliferation
          positive regulation of ATP biosynthetic process
          negative regulation of neuron migration
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          RNA polymerase II repressing transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          signal transducer activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          transcription factor binding
          protein kinase binding
          protein phosphatase binding
          CCR5 chemokine receptor binding
          glucocorticoid receptor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0