Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012772.1 | NP_036904.1 | 1 | 142 |
Gene Symbol: LOC103694545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773683.1 | XP_008771905.1 | 1 | 267 |
Gene Symbol: LOC103694550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773684.1 | XP_008771906.1 | 1 | 303 |