Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012799.1 | NP_036931.1 | 2 | 881 | ||
GenBank mRNA | U37058.1 | - | 2 | 881 |
Set Membership: |
![]() |