Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDGFRA SIRNA Assays ›
          Gene Symbol:
          Pdgfra
          Gene Name:
          platelet derived growth factor receptor, alpha polypeptide
          Gene Aliases:
          APDGFR, PDGFACE
          Chromosome Location:
          Chr.14: 35527926 - 35581129 on Build Rnor_6.0
          UniGene ID:
          Rn.55127
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s129322

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pdgfra
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012802.1 NP_036934.1 21 2934
          XM_006250879.2 XP_006250941.1 21 2989
          XM_006250880.2 XP_006250942.1 22 3295
          XM_006250881.2 XP_006250943.1 21 2943
          GenBank mRNA CB801421.1 - 2 108
          M63837.1 - 21 2939

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          luteinization
          in utero embryonic development
          hematopoietic progenitor cell differentiation
          positive regulation of cytosolic calcium ion concentration
          estrogen metabolic process
          positive regulation of cell proliferation
          female gonad development
          anatomical structure morphogenesis
          response to hormone
          organ morphogenesis
          response to organic substance
          response to inorganic substance
          negative regulation of platelet activation
          positive regulation of phospholipase C activity
          cell migration
          peptidyl-tyrosine phosphorylation
          signal transduction involved in regulation of gene expression
          extracellular matrix organization
          lung development
          adrenal gland development
          positive regulation of cell migration
          male genitalia development
          response to estradiol
          Leydig cell differentiation
          response to cytokine
          platelet-derived growth factor receptor-alpha signaling pathway
          cellular response to platelet-derived growth factor stimulus
          vascular endothelial growth factor signaling pathway
          positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
          wound healing
          gliogenesis
          odontogenesis of dentin-containing tooth
          positive regulation of phosphatidylinositol 3-kinase activity
          response to estrogen
          positive regulation of DNA replication
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          embryonic digestive tract morphogenesis
          embryonic cranial skeleton morphogenesis
          embryonic skeletal system morphogenesis
          skeletal system morphogenesis
          inner ear development
          regulation of chemotaxis
          cardiac myofibril assembly
          response to hyperoxia
          palate development
          face morphogenesis
          cell chemotaxis
          retina vasculature development in camera-type eye
          positive regulation of ERK1 and ERK2 cascade
          platelet aggregation
          cellular response to amino acid stimulus
          metanephric glomerular capillary formation
          regulation of mesenchymal stem cell differentiation
          transmembrane receptor protein tyrosine kinase activity
          platelet-derived growth factor alpha-receptor activity
          vascular endothelial growth factor-activated receptor activity
          platelet-derived growth factor receptor binding
          ATP binding
          protein complex binding
          vascular endothelial growth factor binding
          protein homodimerization activity
          phosphatidylinositol 3-kinase binding
          protein dimerization activity
          platelet-derived growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix