Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012899.2 | NP_037031.1 | 7, 8 | 742 | ||
XM_006238234.2 | XP_006238296.1 | 7, 8 | 1335 | |||
XM_006238235.1 | XP_006238297.1 | 7, 8 | 832 | |||
GenBank mRNA | BC061806.1 | - | 7, 8 | 742 | 5598340 | |
FQ210250.1 | - | 7, 8 | 687 | |||
FQ213585.1 | - | 7, 8 | 884 | |||
FQ218546.1 | - | 7, 8 | 687 | |||
FQ219571.1 | - | 7, 8 | 687 | |||
FQ231154.1 | - | 7, 8 | 718 | |||
FQ232546.1 | - | 7, 8 | 718 | |||
FQ233724.1 | - | 7, 8 | 768 | |||
FQ234245.1 | - | 7, 8 | 718 | |||
FQ234573.1 | - | 7, 8 | 766 | |||
X04959.1 | - | 6, 7 | 624 |