Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012927.1 | NP_037059.1 | 3 | 434 | ||
GenBank mRNA | D25290.1 | - | 3 | 434 |