Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012984.3 | NP_037116.2 | 14, 15 | 2373 | ||
XM_006252830.2 | XP_006252892.1 | 14, 15 | 2412 | |||
XM_006252831.2 | XP_006252893.1 | 14, 15 | 2409 | |||
XM_006252832.2 | XP_006252894.1 | 14, 15 | 2412 | |||
XM_006252833.2 | XP_006252895.1 | 14, 15 | 2409 | |||
XM_006252835.2 | XP_006252897.1 | 14, 15 | 2409 | |||
XM_006252836.2 | XP_006252898.1 | 14, 15 | 2412 | |||
XM_006252837.2 | XP_006252899.1 | 14, 15 | 2409 |