Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4hb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012998.1 | NP_037130.1 | 9, 10 | 1410 | ||
GenBank mRNA | BC061857.1 | - | 9, 10 | 1424 | 5623688 | |
FQ220001.1 | - | 1, 2 | 197 | |||
M21018.1 | - | 9, 10 | 1288 | |||
X02918.1 | - | 9, 10 | 1410 |