Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145366.1 | NP_001138838.1 | 6 | 904 | ||
NM_001145367.1 | NP_001138839.1 | 6 | 1082 | |||
NM_013124.3 | NP_037256.1 | 5 | 879 | |||
XM_006237009.2 | XP_006237071.1 | 6 | 875 | |||
GenBank mRNA | AB011365.1 | - | 4 | 712 | ||
AB019561.1 | - | 5 | 793 | |||
AF156665.1 | - | 6 | 904 | |||
AF156666.1 | - | 5 | 841 | |||
AF246457.1 | - | 6 | 832 | |||
AF246458.1 | - | 6 | 872 | |||
Y12882.2 | - | 5 | 842 |