Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013176.2 | NP_037308.1 | 16 | 1644 | ||
XM_006243345.1 | XP_006243407.1 | 16 | 1499 | |||
XM_006243346.1 | XP_006243408.1 | 16 | 1499 | |||
XM_006243347.1 | XP_006243409.1 | 15 | 1430 | |||
XM_006243348.1 | XP_006243410.1 | 15 | 1427 | |||
XM_006243349.1 | XP_006243411.1 | 15 | 1427 | |||
XM_006243350.1 | XP_006243412.1 | 15 | 1424 | |||
XM_006243351.1 | XP_006243413.1 | 14 | 1355 | |||
XM_006243352.2 | XP_006243414.1 | 9 | 1293 | |||
XM_006243353.2 | XP_006243415.1 | 9 | 1290 | |||
XM_006243354.2 | XP_006243416.1 | 8 | 1218 | |||
XM_008766218.1 | XP_008764440.1 | 14 | 5183 | |||
XM_008766219.1 | XP_008764441.1 | 8 | 1221 | |||
GenBank mRNA | AJ973226.1 | - | 9 | 1018 | ||
L09656.1 | - | 16 | 1644 |