Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022379.1 | NP_071774.1 | 4, 5 | 554 | ||
XM_006236128.2 | XP_006236190.1 | 3, 4 | 820 | |||
GenBank mRNA | L08812.1 | - | 4, 5 | 554 |