Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017182.2 | NP_058878.1 | 6 | 793 | ||
XM_006253573.2 | XP_006253635.1 | 6 | 831 | |||
XM_006253574.2 | XP_006253636.1 | 6 | 921 | |||
XM_006253577.2 | XP_006253639.1 | 6 | 831 | |||
GenBank mRNA | BC089093.1 | - | 6 | 793 | 7126329 | |
M99066.1 | - | 2 | 125 | |||
U79139.1 | - | 6 | 738 |