Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017196.3 | NP_058892.1 | 5, 6 | 456 | ||
XM_006256061.2 | XP_006256123.1 | 4, 5 | 830 | |||
XM_006256062.2 | XP_006256124.1 | 5, 6 | 807 | |||
XM_006256063.2 | XP_006256125.1 | 5, 6 | 770 | |||
XM_006256065.2 | XP_006256127.1 | 7, 8 | 606 | |||
GenBank mRNA | AB000818.1 | - | 5, 6 | 454 | ||
AF299328.1 | - | 2, 3 | 196 | |||
BC081822.1 | - | 3, 4 | 1303 | |||
D82069.1 | - | 7, 8 | 611 | |||
FM053942.1 | - | 5, 6 | 443 | |||
FQ231499.1 | - | 5, 6 | 454 | |||
U10894.1 | - | 2, 3 | 210 | |||
U17919.1 | - | 5, 6 | 428 | |||
U33471.1 | - | 3, 4 | 1480 |