Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019621.1 | NP_062567.1 | 6 | 413 | ||
XM_006246680.2 | XP_006246742.1 | 8 | 789 | |||
XM_006246681.2 | XP_006246743.1 | 8 | 784 | |||
XM_006246682.2 | XP_006246744.1 | 8 | 638 | |||
XM_006246683.2 | XP_006246745.1 | 8 | 546 | |||
XM_006246684.2 | XP_006246746.1 | 8 | 537 | |||
XM_006246685.2 | XP_006246747.1 | 6 | 499 | |||
XM_006246686.2 | XP_006246748.1 | 6 | 559 | |||
XM_006246687.2 | XP_006246749.1 | 6 | 459 | |||
XM_006246688.2 | XP_006246750.1 | 5 | 2660 | |||
XM_006246689.2 | XP_006246751.1 | 5 | 402 | |||
XM_006246691.2 | XP_006246753.1 | 5 | 393 | |||
XM_008767809.1 | XP_008766031.1 | 8 | 890 | |||
XM_008767810.1 | XP_008766032.1 | 5 | 277 | |||
GenBank mRNA | M96853.1 | - | 6 | 413 | ||
X66474.1 | - | 6 | 558 |