Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017221.1 | NP_058917.1 | 1 | 532 | ||
GenBank mRNA | L27340.1 | - | 1 | 532 |