Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022584.2 | NP_072106.1 | 6 | 476 | ||
GenBank mRNA | AF072865.1 | - | 6 | 497 | ||
BC085734.1 | - | 6 | 476 | 7191992 |