Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017327.1 | NP_059023.1 | 1 | 758 | ||
XM_008772320.1 | XP_008770542.1 | 1 | 767 | |||
XM_008772321.1 | XP_008770543.1 | 1 | 767 | |||
GenBank mRNA | M17526.1 | - | 1 | 758 |