Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK1 SIRNA Assays ›
          Gene Symbol:
          Cdk1
          Gene Name:
          cyclin-dependent kinase 1
          Gene Aliases:
          Cdc2, Cdc2a
          Chromosome Location:
          Chr.20: 20576343 - 20591510 on Build Rnor_6.0
          UniGene ID:
          Rn.6934
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s132430

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cdk1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019296.1 NP_062169.1 4 332
          XM_006256353.2 XP_006256415.1 5 459
          GenBank mRNA BC091549.1 - 5 417 7388690
          FQ219810.1 - 4 335
          FQ227059.1 - 4 335
          X60767.1 - 4 332

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          protein complex assembly
          protein phosphorylation
          apoptotic process
          mitotic nuclear division
          mitotic G2 DNA damage checkpoint
          cell aging
          response to toxic substance
          response to organonitrogen compound
          positive regulation of gene expression
          response to organic cyclic compound
          response to amine
          response to activity
          histone phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          chromosome condensation
          epithelial cell differentiation
          organ regeneration
          positive regulation of protein import into nucleus, translocation
          protein localization to kinetochore
          response to drug
          response to hydrogen peroxide
          negative regulation of apoptotic process
          response to ethanol
          positive regulation of DNA replication
          positive regulation of mitotic cell cycle
          response to cadmium ion
          response to copper ion
          response to axon injury
          cell division
          ventricular cardiac muscle cell development
          positive regulation of cardiac muscle cell proliferation
          cellular response to hydrogen peroxide
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding
          RNA polymerase II carboxy-terminal domain kinase activity
          kinase activity
          cyclin binding
          Hsp70 protein binding
          histone kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0