Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019300.1 | NP_062173.1 | 11 | 1211 | ||
GenBank mRNA | U67914.1 | - | 11 | 1197 |