Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AMHR2 SIRNA Assays ›
          Gene Symbol:
          AMHR2
          Gene Name:
          anti-Mullerian hormone receptor type 2
          Gene Aliases:
          AMHR, MISR2, MISRII, MRII
          Chromosome Location:
          Chr.12: 53423855 - 53431672 on Build GRCh38
          UniGene ID:
          Hs.659889
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1325

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AMHR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164690.1 NP_001158162.1 6 850
          NM_001164691.1 NP_001158163.1 6 850
          NM_020547.2 NP_065434.1 6 850
          XM_011538173.1 XP_011536475.1 6 853
          XM_011538174.1 XP_011536476.1 6 850
          XM_011538176.1 XP_011536478.1 6 853
          XM_011538178.1 XP_011536480.1 6 634
          XM_011538179.1 XP_011536481.1 6 853
          XM_011538180.1 XP_011536482.1 6 767
          XM_011538181.1 XP_011536483.1 6 764
          XM_011538183.2 XP_011536485.1 6 853
          XM_011538184.2 XP_011536486.1 6 853
          XM_011538185.1 XP_011536487.1 6 853
          XM_011538186.2 XP_011536488.1 3 162
          XM_011538187.2 XP_011536489.1 6 853
          XM_017019179.1 XP_016874668.1 6 853
          GenBank mRNA AF172932.1 - 6 833
          AK313593.1 - 6 839
          AY714878.1 - 6 770
          BC126316.1 - 6 825 8992032
          BC136356.1 - 6 825 9020343
          CR627318.1 - 3 268

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          Mullerian duct regression
          signal transduction
          transforming growth factor beta receptor signaling pathway
          sex differentiation
          male gonad development
          female gonad development
          signal transduction by protein phosphorylation
          negative regulation of anti-Mullerian hormone signaling pathway
          anti-Mullerian hormone signaling pathway
          receptor signaling protein serine/threonine kinase activity
          receptor activity
          transforming growth factor beta receptor activity, type II
          protein binding
          ATP binding
          hormone binding
          metal ion binding
          anti-Mullerian hormone receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0