Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019362.1 | NP_062235.1 | 15 | 1837 | ||
XM_008761922.1 | XP_008760144.1 | 15 | 1581 | |||
GenBank mRNA | AF068261.1 | - | 15 | 1602 | ||
AF099990.1 | - | 15 | 1541 | |||
D88190.1 | - | 15 | 1837 |