Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYL1 SIRNA Assays ›
Gene Symbol:
Myl1
Gene Name:
myosin, light chain 1
Gene Aliases:
MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
Chromosome Location:
Chr.9: 73937820 - 73958480 on Build Rnor_6.0
UniGene ID:
Rn.84920
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s132759
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077656.1
NP_001071124.1
3
317
NM_020104.2
NP_064489.1
3
135
XM_008767294.1
XP_008765516.1
2
265
XM_008767295.1
XP_008765517.1
2
386
GenBank mRNA
CR755322.1
-
3
140
CR756356.1
-
3
135
FQ214580.1
-
3
444
FQ214604.1
-
3
362
FQ214608.1
-
3
322
FQ214631.1
-
3
304
FQ214638.1
-
3
411
FQ214656.1
-
3
153
FQ214675.1
-
3
411
FQ214712.1
-
3
325
FQ214723.1
-
3
325
FQ214727.1
-
3
325
FQ214742.1
-
3
325
FQ214939.1
-
3
322
FQ215002.1
-
3
411
FQ215050.1
-
3
325
FQ215083.1
-
3
322
FQ215201.1
-
3
322
FQ215229.1
-
3
416
FQ215325.1
-
3
411
FQ215340.1
-
3
153
FQ215418.1
-
3
411
FQ215435.1
-
3
322
FQ215492.1
-
3
304
FQ215500.1
-
2
901
FQ215535.1
-
2
386
FQ215557.1
-
3
411
FQ215563.1
-
3
322
FQ215578.1
-
3
411
FQ215610.1
-
3
325
FQ215748.1
-
2
384
FQ215766.1
-
3
411
FQ215774.1
-
3
322
FQ215820.1
-
3
380
FQ215848.1
-
3
322
FQ215875.1
-
3
322
FQ215911.1
-
3
416
FQ215933.1
-
3
325
FQ216034.1
-
3
325
FQ216060.1
-
3
411
FQ216109.1
-
3
322
FQ216124.1
-
3
416
FQ216181.1
-
3
304
FQ216213.1
-
4
319
FQ216218.1
-
3
322
FQ216229.1
-
3
592
FQ216238.1
-
3
411
FQ216308.1
-
3
153
FQ216407.1
-
3
493
FQ216452.1
-
3
322
FQ216455.1
-
3
326
FQ216484.1
-
3
304
FQ216496.1
-
3
325
FQ216525.1
-
3
325
FQ216543.1
-
3
322
FQ216556.1
-
3
411
FQ216567.1
-
3
322
FQ216631.1
-
3
325
FQ216654.1
-
3
409
FQ216704.1
-
3
325
FQ216737.1
-
3
408
FQ216738.1
-
3
153
FQ217074.1
-
3
152
FQ217103.1
-
3
153
FQ217157.1
-
2
128
FQ217192.1
-
3
153
FQ217218.1
-
3
153
FQ217243.1
-
3
152
FQ217303.1
-
3
304
FQ217323.1
-
3
153
FQ217387.1
-
3
153
FQ217394.1
-
3
255
FQ217427.1
-
3
158
FQ217481.1
-
3
153
FQ217524.1
-
3
153
FQ217753.1
-
3
158
FQ217756.1
-
3
153
FQ217889.1
-
3
153
FQ217973.1
-
3
158
FQ217979.1
-
3
153
FQ218029.1
-
3
153
FQ218049.1
-
3
153
FQ223584.1
-
3
411
FQ223632.1
-
3
325
FQ223641.1
-
3
411
FQ223687.1
-
3
153
FQ223779.1
-
3
153
FQ223827.1
-
3
325
FQ223880.1
-
3
223
FQ223984.1
-
3
153
FQ224047.1
-
3
153
FQ224050.1
-
3
153
FQ224087.1
-
3
153
FQ224112.1
-
2
128
FQ224155.1
-
3
158
FQ224171.1
-
3
322
FQ224172.1
-
3
153
FQ224254.1
-
3
153
FQ224277.1
-
3
325
FQ224321.1
-
3
157
FQ224386.1
-
3
158
FQ224411.1
-
3
153
FQ224420.1
-
3
153
FQ224452.1
-
3
228
FQ224519.1
-
3
322
FQ224609.1
-
4
181
FQ224663.1
-
3
153
FQ224726.1
-
3
255
FQ224796.1
-
3
153
FQ224799.1
-
3
158
FQ224927.1
-
3
411
FQ224931.1
-
3
322
FQ224934.1
-
3
320
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021285, NM_001113387, BC059087, AK003182, AK009938, and others.
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cardiac muscle contraction
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0