Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031627.2 | NP_113815.2 | 9 | 1220 | ||
XM_006234533.2 | XP_006234595.1 | 10 | 1398 | |||
XM_006234534.1 | XP_006234596.1 | 10 | 1339 | |||
XM_006234535.2 | XP_006234597.1 | 10 | 1540 | |||
XM_006234537.1 | XP_006234599.1 | 9 | 1010 | |||
XM_008762013.1 | XP_008760235.1 | 9 | 1130 | |||
GenBank mRNA | BC088750.1 | - | 9 | 1220 | 7314121 | |
U11685.1 | - | 9 | 1227 |