Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCND1 SIRNA Assays ›
          Gene Symbol:
          Ccnd1
          Gene Name:
          cyclin D1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 218090750 - 218100447 on Build Rnor_6.0
          UniGene ID:
          Rn.22279
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s132940

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ccnd1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_171992.4 NP_741989.3 5 877
          XM_008760168.1 XP_008758390.1 5 1118
          GenBank mRNA AB256048.1 - 5 929
          AM258963.2 - 5 930
          D14014.1 - 5 877
          FQ231654.1 - 5 890
          X75207.1 - 5 904

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_007631, BC044841, AK005352, AK170833, M64403, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          re-entry into mitotic cell cycle
          liver development
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          protein phosphorylation
          cellular response to DNA damage stimulus
          lactation
          positive regulation of cell proliferation
          response to organic substance
          response to iron ion
          response to X-ray
          response to organonitrogen compound
          positive regulation of G2/M transition of mitotic cell cycle
          response to organic cyclic compound
          Wnt signaling pathway
          negative regulation of Wnt signaling pathway
          negative regulation of epithelial cell differentiation
          endoplasmic reticulum unfolded protein response
          organ regeneration
          mitotic G1 DNA damage checkpoint
          response to magnesium ion
          response to estradiol
          response to vitamin E
          Leydig cell differentiation
          mammary gland epithelial cell proliferation
          positive regulation of mammary gland epithelial cell proliferation
          response to drug
          response to estrogen
          fat cell differentiation
          response to ethanol
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          regulation of protein kinase activity
          response to steroid hormone
          cell division
          response to glucocorticoid
          response to corticosterone
          response to calcium ion
          regulation of cell cycle
          canonical Wnt signaling pathway
          mammary gland alveolus development
          response to UV-A
          negative regulation of cell cycle arrest
          cellular response to organic substance
          regulation of G1/S transition of mitotic cell cycle
          transcription corepressor activity
          protein kinase activity
          transcription factor binding
          kinase activity
          cyclin-dependent protein serine/threonine kinase regulator activity
          enzyme binding
          protein kinase binding
          protein complex binding
          histone deacetylase binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0