Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MB SIRNA Assays ›
          Gene Symbol:
          Mb
          Gene Name:
          myoglobin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 118101633 - 118108864 on Build Rnor_6.0
          UniGene ID:
          Rn.40511
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s133116

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021588.2 NP_067599.1 3 444
          GenBank mRNA AF197916.1 - 3 417
          BC070511.1 - 3 444 7103504
          FQ214690.1 - 3 476
          FQ214898.1 - 3 475
          FQ214986.1 - 3 476
          FQ215026.1 - 3 476
          FQ215032.1 - 3 475
          FQ215043.1 - 3 475
          FQ215217.1 - 3 474
          FQ215511.1 - 3 474
          FQ215553.1 - 3 475
          FQ215560.1 - 3 474
          FQ215564.1 - 3 474
          FQ215642.1 - 3 485
          FQ215802.1 - 3 476
          FQ215880.1 - 3 475
          FQ215982.1 - 3 475
          FQ216001.1 - 3 474
          FQ216029.1 - 3 473
          FQ216127.1 - 3 473
          FQ216280.1 - 3 482
          FQ216299.1 - 3 476
          FQ216306.1 - 3 477
          FQ216335.1 - 3 476
          FQ216380.1 - 3 475
          FQ216443.1 - 3 474
          FQ216504.1 - 3 475
          FQ216709.1 - 3 475
          FQ216731.1 - 3 474
          FQ216841.1 - 3 473
          FQ217450.1 - 3 475
          FQ217496.1 - 3 474
          FQ218027.1 - 3 475
          FQ223624.1 - 3 474
          FQ223672.1 - 3 471
          FQ223774.1 - 3 476
          FQ223938.1 - 3 476
          FQ224156.1 - 3 472
          FQ224523.1 - 3 475
          FQ224856.1 - 3 475
          FQ224909.1 - 3 475
          FQ224921.1 - 3 472
          FQ224975.1 - 3 475

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          heart development
          response to hormone
          oxygen transport
          slow-twitch skeletal muscle fiber contraction
          response to hydrogen peroxide
          enucleate erythrocyte differentiation
          brown fat cell differentiation
          oxygen transporter activity
          iron ion binding
          oxygen binding
          heme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix