Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022269.2 | NP_071605.2 | 2 | 323 | ||
XM_006249717.2 | XP_006249779.1 | 2 | 345 | |||
XM_006249719.2 | XP_006249781.1 | 2 | 343 | |||
XM_006249723.2 | XP_006249785.1 | 2 | 351 | |||
XM_008769497.1 | XP_008767719.1 | 2 | 345 | |||
XM_008769498.1 | XP_008767720.1 | 2 | 345 | |||
GenBank mRNA | AB026900.1 | - | 2 | 330 | ||
AB026901.1 | - | 2 | 267 | |||
AB026902.1 | - | 2 | 330 | |||
AB026903.1 | - | 2 | 267 | |||
AB026904.1 | - | 1 | 116 | |||
AF039583.1 | - | 2 | 264 | |||
AF039584.1 | - | 2 | 264 | |||
BC061869.1 | - | 2 | 287 | 5598916 | ||
CB610207.1 | - | 2 | 323 |