Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022639.1 | NP_072161.1 | 2 | 219 | ||
XM_006229891.2 | XP_006229953.1 | 1 | 310 | |||
GenBank mRNA | AF196344.1 | - | 2 | 219 |