Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022699.3 | NP_073190.1 | 3 | 120 | ||
GenBank mRNA | BC058471.1 | - | 3 | 124 | 6919868 | |
FQ210232.1 | - | 3 | 117 | |||
FQ210454.1 | - | 3 | 116 | |||
FQ210629.1 | - | 3 | 120 | |||
FQ211005.1 | - | 3 | 122 | |||
FQ211694.1 | - | 3 | 117 | |||
FQ211731.1 | - | 3 | 116 | |||
FQ211953.1 | - | 3 | 120 | |||
FQ214466.1 | - | 3 | 117 | |||
FQ217830.1 | - | 3 | 117 | |||
FQ217913.1 | - | 3 | 117 | |||
FQ221046.1 | - | 3 | 117 | |||
FQ221166.1 | - | 3 | 117 | |||
FQ221653.1 | - | 3 | 125 | |||
FQ221662.1 | - | 3 | 118 | |||
FQ221710.1 | - | 3 | 117 | |||
FQ221740.1 | - | 3 | 116 | |||
FQ221836.1 | - | 3 | 117 | |||
FQ222551.1 | - | 3 | 117 | |||
FQ222688.1 | - | 3 | 117 | |||
FQ222703.1 | - | 3 | 116 | |||
FQ223191.1 | - | 3 | 116 | |||
FQ223319.1 | - | 3 | 117 | |||
FQ223321.1 | - | 3 | 98 | |||
FQ223373.1 | - | 3 | 116 | |||
FQ223410.1 | - | 3 | 214 | |||
FQ223428.1 | - | 3 | 121 | |||
FQ223661.1 | - | 3 | 117 | |||
FQ224433.1 | - | 3 | 119 | |||
FQ224506.1 | - | 3 | 116 | |||
FQ224779.1 | - | 3 | 117 | |||
FQ228655.1 | - | 3 | 116 | |||
FQ228991.1 | - | 3 | 116 | |||
FQ229525.1 | - | 3 | 115 | |||
FQ229603.1 | - | 717 |