Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024147.1 | NP_077061.1 | 2 | 320 | ||
XM_008764850.1 | XP_008763072.1 | 2 | 269 | |||
XM_008764851.1 | XP_008763073.1 | 2 | 2152 | |||
XM_008764852.1 | XP_008763074.1 | 3 | 2117 | |||
XM_008764853.1 | XP_008763075.1 | 3 | 2117 | |||
XM_008764854.1 | XP_008763076.1 | 3 | 2082 | |||
XM_008764855.1 | XP_008763077.1 | 2 | 269 | |||
XM_008764856.1 | XP_008763078.1 | 2 | 420 | |||
XM_008764857.1 | XP_008763079.1 | 2 | 247 | |||
GenBank mRNA | U70211.1 | - | 2 | 320 |