Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024398.2 | NP_077374.2 | 11 | 1323 | ||
GenBank mRNA | AJ243266.1 | - | 11 | 1324 | ||
BC061999.1 | - | 11 | 1323 | 5621226 |