Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PAX8 SIRNA Assays ›
          Gene Symbol:
          Pax8
          Gene Name:
          paired box 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 1527316 - 1586023 on Build Rnor_6.0
          UniGene ID:
          Rn.10392
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s135818

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pax8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031141.2 NP_112403.2 6, 7 763
          XM_006233646.2 XP_006233708.1 6, 7 1863
          XM_008761610.1 XP_008759832.1 5, 6 2199
          XM_008761611.1 XP_008759833.1 5, 6 2200
          XM_008761612.1 XP_008759834.1 3, 4 1927
          XM_008761613.1 XP_008759835.1 5, 6 2195
          GenBank mRNA BC081779.1 - 6, 7 763 7127710

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_011040, X57487, AK050717, BC020526, AK052610.

          Gene Ontology Categories:

          Function(s) Process(es)

          urogenital system development
          metanephros development
          branching involved in ureteric bud morphogenesis
          kidney development
          mesonephros development
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          sulfur compound metabolic process
          central nervous system development
          cell differentiation
          thyroid gland development
          pronephric field specification
          inner ear morphogenesis
          regulation of apoptotic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          pronephros development
          cellular response to gonadotropin stimulus
          otic vesicle development
          positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          metanephric epithelium development
          metanephric distal convoluted tubule development
          metanephric comma-shaped body morphogenesis
          metanephric S-shaped body morphogenesis
          metanephric nephron tubule formation
          negative regulation of mesenchymal cell apoptotic process involved in metanephric nephron morphogenesis
          regulation of metanephric nephron tubule epithelial cell differentiation
          positive regulation of branching involved in ureteric bud morphogenesis
          negative regulation of mesenchymal cell apoptotic process involved in metanephros development
          negative regulation of apoptotic process involved in metanephric collecting duct development
          negative regulation of apoptotic process involved in metanephric nephron tubule development
          positive regulation of metanephric DCT cell differentiation
          positive regulation of thyroid hormone generation
          regulation of thyroid-stimulating hormone secretion
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0