Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nefl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031783.1 | NP_113971.1 | 2, 3 | 1241 | ||
GenBank mRNA | AF031880.1 | - | 2, 3 | 1241 | ||
M25638.1 | - | 2, 3 | 579 |