Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fetub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053348.1 | NP_445800.1 | 7 | 943 | ||
XM_006248549.1 | XP_006248611.1 | 8 | 897 | |||
XM_006248550.1 | XP_006248612.1 | 8 | 1260 | |||
GenBank mRNA | AJ242926.1 | - | 7 | 943 | ||
BC070508.1 | - | 8 | 882 | 7099154 | ||
FQ209449.1 | - | 8 | 880 | |||
FQ209518.1 | - | 7 | 894 | |||
FQ209610.1 | - | 7 | 930 | |||
FQ209832.1 | - | 7 | 895 | |||
FQ210335.1 | - | 7 | 895 | |||
FQ211050.1 | - | 7 | 894 | |||
FQ218557.1 | - | 8 | 876 | |||
FQ218709.1 | - | 7 | 895 | |||
FQ218884.1 | - | 8 | 865 | |||
FQ218893.1 | - | 6 | 795 | |||
FQ218905.1 | - | 7 | 894 | |||
FQ218930.1 | - | 7 | 927 | |||
FQ218949.1 | - | 7 | 744 | |||
FQ219048.1 | - | 7 | 925 | |||
FQ219152.1 | - | 7 | 931 | |||
FQ219170.1 | - | 7 | 894 | |||
FQ219190.1 | - | 7 | 930 | |||
FQ219266.1 | - | 7 | 894 | |||
FQ219352.1 | - | 7 | 894 | |||
FQ219426.1 | - | 5 | 496 | |||
FQ219459.1 | - | 7 | 893 | |||
FQ219500.1 | - | 7 | 897 | |||
FQ219513.1 | - | 7 | 966 |